Learn More
Electron Microscopy Sciences Veco Square Mesh Handle 150 mesh Nickel Grid
SDP
Catalog No. 5028916
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Nickel
Pitch: 167 µm
Hole: 117 µm
Bar: 50 µm
Catalog No. 50-289-16 Supplier Electron Microscopy Sciences Supplier No. HD150NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Nickel
Pitch: 167 µm
Hole: 117 µm
Bar: 50 µm