Learn More
Electron Microscopy Sciences Veco Square Mesh Handle 200 mesh Copper Grid
SDP
Catalog No. 5028923
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm
Catalog No. 50-289-23 Supplier Electron Microscopy Sciences Supplier No. HD200CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm