Learn More
Grainger Square Head Plug Forged Steel 1 1/2 in
Catalog No. 502749587
Encompass_Preferred
The Anvil Square Head Plug is a forged steel plug with a square head drive. It is made in the USA and has a black oxide finish. The plug has coarse threads and is designed for use in patching holes, filling gaps, or plugging pipe. It is available in sizes ranging from 1/4" to 1" diameter with lengths from 1" to 6". The plugs meet ASME B18.2.1 standards for square drive dimensions. They offer a cost-effective plugging solution for industrial applications.
Catalog No. 50-274-9587 Supplier Grainger Supplier No. 14J747
The Anvil Square Head Plug is a forged steel plug with a square head drive. It is made in the USA and has a black oxide finish. The plug has coarse threads and is designed for use in patching holes, filling gaps, or plugging pipe. It is available in sizes ranging from 1/4" to 1" diameter with lengths from 1" to 6". The plugs meet ASME B18.2.1 standards for square drive dimensions. They offer a cost-effective plugging solution for industrial applications.