Learn More
Grainger Square Head Plug Forged Steel 3/4 in
Catalog No. 502749586
Encompass_Preferred
The Anvil Square Head Plug is a forged steel plug with a square head drive. It is made in the USA and has a black oxide finish. The plug has coarse threads and is designed for use in repairing stripped threads or sealing holes. It is available in multiple sizes ranging from 1/4"-20 to 1"-8 threads. The plugs meet ASME B18.2.1 specifications and offer durability and strength for industrial applications. With proper hole preparation, these plugs provide an economical way to repair threads or seal openings.
Catalog No. 50-274-9586 Supplier Grainger Supplier No. 14J745
The Anvil Square Head Plug is a forged steel plug with a square head drive. It is made in the USA and has a black oxide finish. The plug has coarse threads and is designed for use in repairing stripped threads or sealing holes. It is available in multiple sizes ranging from 1/4"-20 to 1"-8 threads. The plugs meet ASME B18.2.1 specifications and offer durability and strength for industrial applications. With proper hole preparation, these plugs provide an economical way to repair threads or seal openings.