Learn More
Electron Microscopy Sciences Square Mesh | 400 mesh | Copper | 25/box
SDP
Catalog No. NC1648302
Encompass_Preferred
A thin film of pure carbon deposited on one side of the grid. Standard thickness: Approx. 5-6nm
Catalog No. NC1648302 Supplier Electron Microscopy Sciences Supplier No. CF400CU
A thin film of pure carbon deposited on one side of the grid. Standard thickness: Approx. 5-6nm