Learn More
Electron Microscopy Sciences Veco Square Center Reference 200 mesh Nickel Grid
SDP
Catalog No. 5028933
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Nickel
Pitch: 50%
Catalog No. 50-289-33 Supplier Electron Microscopy Sciences Supplier No. E200NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Nickel
Pitch: 50%