Learn More
Electron Microscopy Sciences Veco Square Mesh Oyster Grid D75, Copper
SDP
Catalog No. 5028927
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Pitch: 333 µm
Holes: 283 µm
Bar: 50 µm
Catalog No. 50-289-27 Supplier Electron Microscopy Sciences Supplier No. D75CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Pitch: 333 µm
Holes: 283 µm
Bar: 50 µm