Learn More
Andwin Scientific SQUARE PETRI DISH 100/PK
SDP
Catalog No. 50930468
Encompass_Preferred
The square Electron Microscopy Science Petri Dishes are ideal for use in routine procedures and automatic equipments. This product is constructed from polystyrene that permits superior optical clarity. It is sold in a pack of 100.
Catalog No. 50-930-468 Supplier Andwin Scientific Supplier No. 70690
The square Electron Microscopy Science Petri Dishes are ideal for use in routine procedures and automatic equipments. This product is constructed from polystyrene that permits superior optical clarity. It is sold in a pack of 100.