Learn More
Andwin Scientific Square Petri Dish Grid 10x10, 500 pack
SDP
Catalog No. 50727391
Encompass_Preferred
The Square Petri Dish is made to be utilzed for phase typing, susceptibility testing, plate counts, and probe assays. This product is sterile and is free from optical distortion. The dish features a grid design, with each grid measuring 10 x 10 mm. The grids have numerical markings in one direction and alphabetical markings in the other, which both allow easy sample identification. There are also four venting ribs into the underside of the lid to eliminate the chance of condensation build-up. It is sold in a pack of 500.
Catalog No. 50-727-391 Supplier Andwin Scientific Supplier No. 70691CS
The Square Petri Dish is made to be utilzed for phase typing, susceptibility testing, plate counts, and probe assays. This product is sterile and is free from optical distortion. The dish features a grid design, with each grid measuring 10 x 10 mm. The grids have numerical markings in one direction and alphabetical markings in the other, which both allow easy sample identification. There are also four venting ribs into the underside of the lid to eliminate the chance of condensation build-up. It is sold in a pack of 500.