Learn More
Electron Microscopy Sciences Square Petri Dish, Plain
SDP
Catalog No. 501900278
Encompass_Preferred
This polystyrene radiation sterile dish commonly referred to as a 100 x 15 mm dish. Free from optical distortion and sterile, these dishes are ideal for phase typing, susceptibility testing, plate counts, and probe assays.
Catalog No. 50-190-0278 Supplier Electron Microscopy Sciences Supplier No. 70690CS
This polystyrene radiation sterile dish commonly referred to as a 100 x 15 mm dish. Free from optical distortion and sterile, these dishes are ideal for phase typing, susceptibility testing, plate counts, and probe assays.