Learn More
Alkali Scientific ASI™ Square Petri Dish with Grid, PS, Sterile, 243 x 243 x 27.3mm, 20/Case
SDP
Catalog No. 503029260
Encompass_Preferred
The ASI™ Square Petri Dish offers a reliable and efficient solution for microbiological applications, including bacterial culture, fungal growth, and sample storage. Manufactured from high-quality polystyrene (PS), these sterile petri dishes provide excellent optical clarity for easy colony observation. The square design maximizes usable surface area, making them ideal for high-throughput applications and grid-based analysis. Each dish is precision-molded for uniformity and stack ability, ensuring secure handling in laboratory settings. Supplied in a sterile format and packaged with 240 per case, these petri dishes are perfect for research, clinical, and industrial microbiology applications. ▪ Ideal for colony formation and enhancing plating efficiency▪ Useful for counting or colony picking
Catalog No. 50-302-9260 Supplier Alkali Scientific Supplier No. PD0127
The ASI™ Square Petri Dish offers a reliable and efficient solution for microbiological applications, including bacterial culture, fungal growth, and sample storage. Manufactured from high-quality polystyrene (PS), these sterile petri dishes provide excellent optical clarity for easy colony observation. The square design maximizes usable surface area, making them ideal for high-throughput applications and grid-based analysis. Each dish is precision-molded for uniformity and stack ability, ensuring secure handling in laboratory settings. Supplied in a sterile format and packaged with 240 per case, these petri dishes are perfect for research, clinical, and industrial microbiology applications. ▪ Ideal for colony formation and enhancing plating efficiency▪ Useful for counting or colony picking