Learn More
Electron Microscopy Sciences Square Petri Dish with Grid, 10/sleeve, 100/pk
SDP
Catalog No. 50365047
Encompass_Preferred
This polystyrene radiation sterile dish commonly referred to as a 100 x 15 mm dish. Free from optical distortion and sterile, these dishes are ideal for phase typing, susceptibility testing, plate counts, and probe assays. Grid of 36 of 13 mm squares marked numerically in one direction and alphabetically in the other.
Catalog No. 50-365-047 Supplier Electron Microscopy Sciences Supplier No. 70691
This polystyrene radiation sterile dish commonly referred to as a 100 x 15 mm dish. Free from optical distortion and sterile, these dishes are ideal for phase typing, susceptibility testing, plate counts, and probe assays. Grid of 36 of 13 mm squares marked numerically in one direction and alphabetically in the other.