Learn More
Electron Microscopy Sciences Plastic Box Friction-Fitted Lid, 1-11/16" x 1-11/16" x 9/16"
SDP
Catalog No. 5027436
Encompass
The sqare boxes are molded from polystyrene for optimum clarity and rigidity. They have two equal dimensions for length and width. With friction fitting lid.
Square: 1-11/16"
Height: 9/16"
Catalog No. 50-274-36 Supplier Electron Microscopy Sciences Supplier No. 64307
The sqare boxes are molded from polystyrene for optimum clarity and rigidity. They have two equal dimensions for length and width. With friction fitting lid.
Square: 1-11/16"
Height: 9/16"