Learn More
Electron Microscopy Sciences Plastic Box, Hinged Lid Snap Lock, 2-9/16" Square x 2-3/16"
SDP
Catalog No. 5027434
Encompass_Preferred
The sqare boxes are molded from polystyrene for optimum clarity and rigidity. They have two equal dimensions for length and width. With hinged snap-lock lid.
Square: 2-9/16"
Height: 2-3/16"
Lid: 1/4"
Base: 1-15/16"
Catalog No. 50-274-34 Supplier Electron Microscopy Sciences Supplier No. 64313
The sqare boxes are molded from polystyrene for optimum clarity and rigidity. They have two equal dimensions for length and width. With hinged snap-lock lid.
Square: 2-9/16"
Height: 2-3/16"
Lid: 1/4"
Base: 1-15/16"