Learn More
Grainger 80/20 Square Tube Profile, Aluminum, 1 in x 1 in, 145 in, 1 ea
Catalog No. 502786240
Encompass_Preferred
This square Quick Frame profile features a 2.54 cm x 2.54 cm (1 in. x 1 in.) cross-section with a grooved interior designed to securely hold nylon connectors. Constructed from durable aluminum with a smooth, anodized finish, this profile offers a sleek appearance and is easy to clean. It is well-suited for a variety of structural applications including framing, displays, and retail kiosks.
  • Features a press-fit, boltless design
  • Designed for use with the Quick Frame system
  • Smooth surface finish
  • Corrosion-resistant anodized coating
  • Durable aluminum construction
  • Square profile with 1/16 in. wall thickness
Catalog No. 50-278-6240 Supplier Grainger Supplier No. 18G633
This square Quick Frame profile features a 2.54 cm x 2.54 cm (1 in. x 1 in.) cross-section with a grooved interior designed to securely hold nylon connectors. Constructed from durable aluminum with a smooth, anodized finish, this profile offers a sleek appearance and is easy to clean. It is well-suited for a variety of structural applications including framing, displays, and retail kiosks.
  • Features a press-fit, boltless design
  • Designed for use with the Quick Frame system
  • Smooth surface finish
  • Corrosion-resistant anodized coating
  • Durable aluminum construction
  • Square profile with 1/16 in. wall thickness