Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow scientific™ Square Weighing Boats
GREENER_CHOICE

Catalog No. 01677168
Change view
Click to view available options
Bottom Shape:
Flat
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bottom Shape
01-677-168 Flat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-677-168 Supplier Heathrow scientific™ Supplier No. HS1422
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Weigh solid products such as powders, crystals and other small particles. Heathrow Scientific™ Square Weighing Boats feature smooth surfaces to provide minimal loss of sample product.

  • Shallow boat with a wide, flat bottom to resist tipping
  • Ideal for solid product weighing
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • Temperature range -10 to 70°C (14 to 158°F)
  • Can be used with microwave moisture analyzers
  • FDA/food-grade compliant
  • Disposable
  • Pack of 500

Specifications

Bottom Shape Flat
Color White
Disposable Single-use
Material Polystyrene
Shape Square, Flat
Sterility Non-sterile
For Use With (Application) Solid product weighing
Size Medium; 90 x 90 x 7 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.