Learn More
Cole-Parmer Thermo Scientific Nalgene 2110-0006 Square Wide-Mouth Bottle, PPCO, 175 mL, 12/pk
Catalog No. NC1327808 Shop All Cole-Parmer Products
Encompass_Preferred
These space-saving leakproof bottles can withstand temperatures to 275°> F (135°> C). Wide mouth is easy to fill. Shape offers visible label surface so they are ideal as sample mailers. Has leakproof polyproplene (PP) screw closures. Autoclavable.
Catalog No. NC1327808 Supplier Cole-Parmer Supplier No. 0602262
These space-saving leakproof bottles can withstand temperatures to 275°> F (135°> C). Wide mouth is easy to fill. Shape offers visible label surface so they are ideal as sample mailers. Has leakproof polyproplene (PP) screw closures. Autoclavable.