Learn More
Grainger 3M Squeak Reduction Tape 5430, UHMW Polyethylene, Transparent, 36 yds
Catalog No. 502755683
Encompass_Preferred
3M™ Squeak Reduction Tape 5430 is a transparent, UHMW polyethylene film tape featuring a high-tack acrylic adhesive. Designed for industrial and automotive use, it effectively controls and eliminates noise, squeaks, and rattles caused by vibration or wear between plastic and metal components. Its low coefficient of friction and excellent abrasion resistance make it suitable for various sliding and wear applications.
  • Features a low coefficient of friction for applications requiring a slick surface
  • Provides high-tack adhesion to many painted metal and plastic surfaces
  • Demonstrates excellent abrasion and chemical resistance
  • Includes a liner ideal for fabrication and die-cutting into various shapes
  • Performs in temperatures from -30°F to 225°F
  • Suitable for indoor and outdoor environments
Catalog No. 50-275-5683 Supplier Grainger Supplier No. 15D196
3M™ Squeak Reduction Tape 5430 is a transparent, UHMW polyethylene film tape featuring a high-tack acrylic adhesive. Designed for industrial and automotive use, it effectively controls and eliminates noise, squeaks, and rattles caused by vibration or wear between plastic and metal components. Its low coefficient of friction and excellent abrasion resistance make it suitable for various sliding and wear applications.
  • Features a low coefficient of friction for applications requiring a slick surface
  • Provides high-tack adhesion to many painted metal and plastic surfaces
  • Demonstrates excellent abrasion and chemical resistance
  • Includes a liner ideal for fabrication and die-cutting into various shapes
  • Performs in temperatures from -30°F to 225°F
  • Suitable for indoor and outdoor environments