Learn More
Grainger 3M Squeak Reduction Tape, UHMW Polyethylene Film, Translucent, 6 in. Width, 36 yd. Length
Catalog No. 502754792
Encompass_Preferred
These squeak reduction tapes feature a translucent, abrasion-resistant UHMW polyethylene film backing coated with a high-tack acrylic adhesive. Designed for the automotive industry, these tapes help control squeaks and rattles caused by the vibration or wear of metal and plastic materials. The tape provides a low coefficient of friction for applications requiring a slick surface and is suitable for both indoor and outdoor use. It is supplied on a densified kraft paper liner, making it ideal for fabrication and die-cutting into various shapes and sizes.
  • Reduces and helps eliminate noise and vibration
  • Abrasion-resistant backing provides a long wear life
  • Excellent adhesion to many painted metal and plastic surfaces
  • Low coefficient of friction for sliding applications
  • Good chemical resistance
  • Easy to die-cut to meet specific application demands
Catalog No. 50-275-4792 Supplier Grainger Supplier No. 15D287
These squeak reduction tapes feature a translucent, abrasion-resistant UHMW polyethylene film backing coated with a high-tack acrylic adhesive. Designed for the automotive industry, these tapes help control squeaks and rattles caused by the vibration or wear of metal and plastic materials. The tape provides a low coefficient of friction for applications requiring a slick surface and is suitable for both indoor and outdoor use. It is supplied on a densified kraft paper liner, making it ideal for fabrication and die-cutting into various shapes and sizes.
  • Reduces and helps eliminate noise and vibration
  • Abrasion-resistant backing provides a long wear life
  • Excellent adhesion to many painted metal and plastic surfaces
  • Low coefficient of friction for sliding applications
  • Good chemical resistance
  • Easy to die-cut to meet specific application demands