Learn More
Grainger 3M Squeak Reduction Tape, UHMW Polyethylene, 6 in Width
Catalog No. 502756001
Encompass_Preferred
3M™ Squeak Reduction Tape 9325 is an ultra-high molecular weight polyethylene (UHMW-PE) film tape coated with a high-tack acrylic adhesive. It is designed to create a low-friction slip plane effect between incompatible surfaces, effectively controlling squeaks and rattles caused by vibration or wear.
  • Features a thin, abrasion-resistant backing that outperforms PTFE in many sliding applications
  • Provides excellent adhesion to a variety of substrates, including metals and various plastics
  • Suitable for automotive instrument panel and interior noise reduction
  • Ideal for fabrication and die-cutting into custom shapes and lengths
  • Maintains performance across a temperature range of -30°F to 225°F (-34°C to 107°C)
  • Ideal for use where traditional lubrication is not possible due to contamination concerns
Catalog No. 50-275-6001 Supplier Grainger Supplier No. 15D289
3M™ Squeak Reduction Tape 9325 is an ultra-high molecular weight polyethylene (UHMW-PE) film tape coated with a high-tack acrylic adhesive. It is designed to create a low-friction slip plane effect between incompatible surfaces, effectively controlling squeaks and rattles caused by vibration or wear.
  • Features a thin, abrasion-resistant backing that outperforms PTFE in many sliding applications
  • Provides excellent adhesion to a variety of substrates, including metals and various plastics
  • Suitable for automotive instrument panel and interior noise reduction
  • Ideal for fabrication and die-cutting into custom shapes and lengths
  • Maintains performance across a temperature range of -30°F to 225°F (-34°C to 107°C)
  • Ideal for use where traditional lubrication is not possible due to contamination concerns