Learn More
Grainger 3M Squeak Reduction Tape, UHMW Polyethylene, Transparent, 5 yd
Catalog No. 502756428
Encompass_Preferred
Squeak Reduction Tape 5430 is a transparent, single-sided UHMW polyethylene film tape engineered to control squeaks and rattles caused by the vibration or wear of metal and plastic parts. Featuring a high-tack acrylic adhesive, this tape provides a strong initial bond and serves as an effective solution for applications where lubrication is not feasible due to contamination risks. Its low coefficient of friction makes it ideal for slick surface requirements, including automotive instrument panels and interiors.
  • Provides excellent abrasion resistance and good chemical resistance for a long wear life
  • Features a liner suitable for fabrication and die-cutting into various shapes and sizes
  • Performs in temperatures ranging from -30°F to 225°F
  • Delivers strong adhesion to many painted metal and plastic surfaces
  • Designed as a continuous roll for versatile industrial use
Catalog No. 50-275-6428 Supplier Grainger Supplier No. 15D199
Squeak Reduction Tape 5430 is a transparent, single-sided UHMW polyethylene film tape engineered to control squeaks and rattles caused by the vibration or wear of metal and plastic parts. Featuring a high-tack acrylic adhesive, this tape provides a strong initial bond and serves as an effective solution for applications where lubrication is not feasible due to contamination risks. Its low coefficient of friction makes it ideal for slick surface requirements, including automotive instrument panels and interiors.
  • Provides excellent abrasion resistance and good chemical resistance for a long wear life
  • Features a liner suitable for fabrication and die-cutting into various shapes and sizes
  • Performs in temperatures ranging from -30°F to 225°F
  • Delivers strong adhesion to many painted metal and plastic surfaces
  • Designed as a continuous roll for versatile industrial use