Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Vikan Table Squeegee, TPE Rubber, 9.6 in, White, 1 ea

Catalog No. 501909440
Encompass

This ultra-hygienic single-bladed squeegee is designed for removing excess water and food debris from food-preparation surfaces. Its soft and flexible blade is fully molded to ensure easy and thorough cleaning, making it ideal for environments where hygiene and food safety are crucial.

  • Fully-molded blade prevents common harborage points and contamination traps
  • Ergonomic design enhances comfort and reduces worker strain
  • Contains food-safe ISCC Certified Bio-Circular Plastic
  • Built-in splash guard to prevent liquids from splashing onto dried surfaces
  • Handheld size ideal for cleaning workbenches and tables
  • Compatible with all Euro threaded handles for secure tool attachment
  • One-piece design enhances durability and reduces harborage points
  • Color-coded for use with hygienic zoning plans

Catalog No. 50-190-9440 Supplier MSC Supplier No. 47167564
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This ultra-hygienic single-bladed squeegee is designed for removing excess water and food debris from food-preparation surfaces. Its soft and flexible blade is fully molded to ensure easy and thorough cleaning, making it ideal for environments where hygiene and food safety are crucial.

  • Fully-molded blade prevents common harborage points and contamination traps
  • Ergonomic design enhances comfort and reduces worker strain
  • Contains food-safe ISCC Certified Bio-Circular Plastic
  • Built-in splash guard to prevent liquids from splashing onto dried surfaces
  • Handheld size ideal for cleaning workbenches and tables
  • Compatible with all Euro threaded handles for secure tool attachment
  • One-piece design enhances durability and reduces harborage points
  • Color-coded for use with hygienic zoning plans

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.