Learn More
Grainger Unger Squeegee Handle, Pro Aluminum, 3.0° Taper, 1 in. Diameter, 1 ea
Catalog No. 502790649
Encompass_Preferred
The Pro Aluminum 3.0° handle is a professional-grade cleaning tool component featuring a 1.40 m (55.12 in.) anodized aluminum construction. Designed for versatility and durability, this handle combines a 3.0° taper with an ACME threaded zinc alloy tip, allowing for secure attachment to a variety of squeegees, including AquaDozer® and WaterWand™ models, as well as tools with universal threaded sockets.
  • Features a 1 in. diameter anodized aluminum handle with a fluted finish for enhanced strength.
  • Equipped with an ACME threaded zinc alloy tip compatible with all ACME threaded products.
  • Includes a convenient hang-up hole for efficient storage.
  • Ergonomic design helps reduce hand fatigue during extended cleaning tasks.
  • Corrosion-resistant construction suitable for demanding commercial and industrial environments.
Catalog No. 50-279-0649 Supplier Grainger Supplier No. 169T25
The Pro Aluminum 3.0° handle is a professional-grade cleaning tool component featuring a 1.40 m (55.12 in.) anodized aluminum construction. Designed for versatility and durability, this handle combines a 3.0° taper with an ACME threaded zinc alloy tip, allowing for secure attachment to a variety of squeegees, including AquaDozer® and WaterWand™ models, as well as tools with universal threaded sockets.
  • Features a 1 in. diameter anodized aluminum handle with a fluted finish for enhanced strength.
  • Equipped with an ACME threaded zinc alloy tip compatible with all ACME threaded products.
  • Includes a convenient hang-up hole for efficient storage.
  • Ergonomic design helps reduce hand fatigue during extended cleaning tasks.
  • Corrosion-resistant construction suitable for demanding commercial and industrial environments.