Learn More
Braintree Scientific Squeekum Rat Model Replacement Tail
SDP
Catalog No. 501955931
Encompass
Rescue Critters' Squeekums Rat manikin allows students, lab technicians, and hWheatons to learn how to confidently handle and safely manage lab rodents.The tail of this amazing rodent model can be conveniently detached and disposed of, while also having the capability of IV access at the caudal vein site. Tiny replaceable ears help students learn ear tagging procedures. The model is fully articulated and extremely realistic. Head, feet, and limbs move in a natural manner. Squeekums comes with its own protective case, IV accessories, artificial blood, and instructions. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5931 Supplier Braintree Scientific Supplier No. SQUEEK2
Rescue Critters' Squeekums Rat manikin allows students, lab technicians, and hWheatons to learn how to confidently handle and safely manage lab rodents.The tail of this amazing rodent model can be conveniently detached and disposed of, while also having the capability of IV access at the caudal vein site. Tiny replaceable ears help students learn ear tagging procedures. The model is fully articulated and extremely realistic. Head, feet, and limbs move in a natural manner. Squeekums comes with its own protective case, IV accessories, artificial blood, and instructions. **Sold for research purposes only and not for human use**