Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3108F(x) Tool Lanyard Single Locking Carabiner

Catalog No. 19910462 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Load Bearing Capacity (English):
15 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity (English)
19-910-462 15 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-910-462 Supplier Ergodyne™ Supplier No. 19808
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Squids™ 3108F(x) Tool Lanyard Single Locking Carabiner is designed to prevent falling objects and reduce shock load.

  • Approved to the ANSI/ISEA 121-2018 standard
  • Shock absorbing properties reduce the dynamic force on the body or other anchor point
  • Durable 1680 denier polyester elastic blended webbing extends
  • Low profile lightweight design reduces snag hazards and added weight
  • Anodized aluminum alloy carabiner with double action locking gate and inherent captive eye
  • Ergonomic carabiner is designed for easily gripping and handling with gloves
  • Treaded loop end that grips the tool when attached
  • High strength 3-ply nylon stitching secures connecting ends
  • Single length construction eliminates weak points seen with multi-component lanyards
  • Certified to a 2:1 safety factor

Specifications

Color Orange, Gray
Length (English) 38 to 48 in.
Length (Metric) 96 to 122 cm
Load Bearing Capacity (English) 15 lb.
Certifications/Compliance ANSI/ISEA 121-2018 standard
Load Bearing Capacity (Metric) 6.8 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.