Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3114 Pull-On Wrist Lanyard with Carabiner

Catalog No. 19820384 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Load Bearing Capacity (Metric):
1.4 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity (Metric)
19-820-384 1.4 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-820-384 Supplier Ergodyne™ Supplier No. 19056
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Squids™ 3114 Pull-On Wrist Lanyard with Carabiner is designed to tether tools and reduces snag hazards.

  • Elastic maintains snug fit
  • Pull-on design is easy to take on and off
  • One size fits all
  • Stainless steel carabiner for easy tool attachment
  • Tested using a 2:1 safety factor

Specifications

Color Black
Material Stainless Steel
Load Bearing Capacity (Metric) 1.4 kg
Certifications/Compliance ANSI/ISEA 121-2018 standard
Load Bearing Capacity (English) 3 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.