Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3130S Coiled Cable Lanyard

Catalog No. 19820386 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Load Bearing Capacity (Metric):
0.9 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity (Metric)
19-820-386 0.9 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-820-386 Supplier Ergodyne™ Supplier No. 19130
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Squids™ 3130S Coiled Cable Lanyard reduces snag hazards of traditional lanyards while preventing dropped objects.

  • Approved to the ANSI/ISEA 121-2018 standard
  • Constructed with PU covered aircraft cable
  • Stainless steel carabiners
  • Coil design prevents snag and tangle hazards
  • Swiveling design prevents tangles and fast recoil
  • Captive eye carabiner
  • Certified to a 2:1 safety factor

Specifications

Color Lime
Material Stainless Steel
Load Bearing Capacity (Metric) 0.9 kg
Length (English) 6.5 to 48.0 in.
Length (Metric) 16.5 to 123 cm
Load Bearing Capacity (English) 2 lb.
Certifications/Compliance ANSI/ISEA 121-2018 standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.