Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3704 Wire Tool Attachment with Loop Tool Tail™

Catalog No. 19820333 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-820-333 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-820-333 Supplier Ergodyne™ Supplier No. 19707
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Squids™ 3704 Wire Tool Attachment with Loop Tool Tail™ features a loop end to securely connect tool lanyards to tools with small captive holes.

  • Stainless steel cable wire is corrosion resistant to withstand weathering
  • PU coating over cable wire for added protection and durability
  • Easily loops to create a one-step attachment point
  • Designed to safety tether small captive holes on tools

Specifications

Color Orange
Length (Metric) 15 cm
Length (English) 6 in.
Includes 6/Pk.
Load Bearing Capacity (English) 2 lb.
Load Bearing Capacity (Metric) 0.9 kg
Certifications/Compliance ANSI/ISEA 121-2018 standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.