Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3193 Tape Measure Tethering Kit - 2lbs/0.9kg

Catalog No. 19820324 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Includes:
Squids™ 3100F(x) Single Carabiner Tool Lanyard and a Squids™ 3770 Tape Measure Trap
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-820-324 Squids™ 3100F(x) Single Carabiner Tool Lanyard and a Squids™ 3770 Tape Measure Trap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-820-324 Supplier Ergodyne™ Supplier No. 19663
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tool Tethering Kit allows users to tether tape measures making it easy to use tools on the job while preventing dropped objects when working at-heights.

  • Tether one standard 2lb / 0.9kg tape measure
  • Straightforward installation
  • Lack of tool lanyards can create a hazardous environment while working at heights
  • No tape and tool tails required
  • Open the hook-and-loop straps on the Tape Measure Trap. Place the tape measure inside of the trap, with the back of the tape measure facing the D-ring on the back of the trap. Adjust and tighten the hook-and-loop straps accordingly. Attach the 3100F(x) Tool Lanyard to complete the tethering system

Specifications

Certifications/Compliance ANSI/ISEA 121-2018
Load Bearing Capacity (English) 2 lb.
Load Bearing Capacity (Metric) 0.9 kg
Includes Squids™ 3100F(x) Single Carabiner Tool Lanyard and a Squids™ 3770 Tape Measure Trap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.