Learn More
Zymo Research Corporation Squisher™-Single (10 Pack)
SDP
Catalog No. 502037201
Encompass_Preferred
Works well with liver and brain tissue as well as mouse tail snips and small insects. Intended for use with conventional style 1.5 ml microcentrifuge tubes.
Catalog No. 50-203-7201 Supplier Zymo Research Corporation Supplier No. H1001
Works well with liver and brain tissue as well as mouse tail snips and small insects. Intended for use with conventional style 1.5 ml microcentrifuge tubes.