Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sqwincher™ Activity Drinks, Powder Concentrate

Catalog No. 19037309
Change view
Click to view available options
Volume (Metric):
0.7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-037-309 0.7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-037-309 Supplier Sqwincher™ Supplier No. 016046
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thirst-quenching drink that's low in sodium and high in potassium

  • Formulated with electrolytes and nutrients
  • Replenishes fluids, minerals, and sugars depleted through physical exertion
  • Prevents or reduces severity of heat-stress disorders
  • Provides supplemental source of energy
  • Conveniently packaged as an instant powder mix (Powder Pack™)

Specifications

Product Type Activity Drink
Flavor Grape
Format Powder Concentrate
For Use With (Application) Provides supplemental source of energy, prevents or reduces severity of heat-stress disorders
Quantity 32 Pk./Cs.
Volume (English) 23.8 oz.
Volume (Metric) 0.7 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.