Learn More
Grainger SQZBTL LUBE W/ TFE 2OZ TRI-FLO
Supplier:  Grainger 3JDL7
Encompass_Preferred
Tri-Flow Lubricants, with PTFE Lube,2OZ, Unique coating action fills in surface irregularities with premium petroleum oils,extreme pressure additives, Low viscosity, Protects against moisture, corrosion damage, stops icing, rust formation
Catalog No. NC9920631