Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SR-1114 1mg | 2769141-82-2 | 830.86 g/mol | C39H42N8O11S | 1 MG
SDP

Catalog No. 5000226972
Encompass_Preferred

SR-1114 is a PROTAC degrader that targets the ENL protein and induces cereblon-dependent degradation, making it useful for research into epigenetic regulation and cancer biology.

  • PROTAC mechanism: induces targeted degradation of ENL
  • Physical form: solid powder for laboratory use
  • Molecular weight: 830.86 g/mol
  • Chemical formula: C39H42N8O11S
  • High purity suitable for research applications
  • Recommended storage: powder -20°C (long term) or 4°C (short term)
  • Available in milligram-scale pack sizes for research use

Catalog No. 50-002-26972 Supplier Medchemexpress LLC Supplier No. HY1158811MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SR-1114 is a PROTAC degrader that targets the ENL protein and induces cereblon-dependent degradation, making it useful for research into epigenetic regulation and cancer biology.

  • PROTAC mechanism: induces targeted degradation of ENL
  • Physical form: solid powder for laboratory use
  • Molecular weight: 830.86 g/mol
  • Chemical formula: C39H42N8O11S
  • High purity suitable for research applications
  • Recommended storage: powder -20°C (long term) or 4°C (short term)
  • Available in milligram-scale pack sizes for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.