Learn More
Medchemexpress LLC SR 11302 | 160162-42-5 | 99.4% | 376.53 g/mol | C26H32O2 | 10 MG
SDP
Catalog No. 5000288331
Encompass_Preferred
SR 11302 is a synthetic retinoid research compound that selectively inhibits activator protein-1 (AP-1) transcriptional activity and is used in cell-based and in vivo studies to probe AP-1-mediated signaling pathways.
  • Selective inhibition of AP-1 transcriptional activity.
  • High purity (99.44%).
  • Molecular weight 376.53 g/mol and formula C26H32O2.
  • Available as powder and solution formats for flexibility in assays.
  • Suitable for biochemical, cellular, and in vivo research applications.
Catalog No. 50-002-88331 Supplier Medchemexpress LLC Supplier No. HY1587010MG
SR 11302 is a synthetic retinoid research compound that selectively inhibits activator protein-1 (AP-1) transcriptional activity and is used in cell-based and in vivo studies to probe AP-1-mediated signaling pathways.
  • Selective inhibition of AP-1 transcriptional activity.
  • High purity (99.44%).
  • Molecular weight 376.53 g/mol and formula C26H32O2.
  • Available as powder and solution formats for flexibility in assays.
  • Suitable for biochemical, cellular, and in vivo research applications.