Learn More
Medchemexpress LLC HY-15870 5mg Medchemexpress, SR 11302 CAS:160162-42-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY158705MG
Encompass_Preferred
Medchemexpress, HY-15870 5mg SR 11302 CAS:160162-42-5 SR 11302 is an activator protein-1 (AP-1) transcription factor inhibitor. SR 11302 is a retinoid that specifically inhibits AP-1 activity without activating the transcription of retinoic acid response element (RARE). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1809