Learn More
Enzo Life Sciences SR-12813 (10mg). CAS: 126411-39-0
SDP
Supplier:  Enzo Life Sciences BMLGR2420010
Encompass_Preferred
Potent and selective PXR agonist. (EC50=200 nM for human PXR). A weak activator of rat and mouse PXR. Inhibits cholesterol biosynthesis in a number of species. Alternative name: Tetraethyl 2-(3,5-Di-t-butyl-4-hydroxyphenyl)ethenyl-1,1-bisphosphonate. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0945