Learn More
Cayman Chemical SR 1555 (hydrochloride), Purity: ≥98%, CAS: 2309312-90-9, 10 mg
SDP
Catalog No. 502954128
Encompass_Preferred
A selective ligand of RORγ (IC50 = 1 μM); does not bind RORα, LXR, or FXR; acts as an inverse agonist of RORγ, inhibiting endogenous IL-17A gene expression and suppressing differentiation of TH17 cells; stimulates T regulatory development
Catalog No. 50-295-4128 Supplier Cayman Chemical Supplier No. 1207110 MG
A selective ligand of RORγ (IC50 = 1 μM); does not bind RORα, LXR, or FXR; acts as an inverse agonist of RORγ, inhibiting endogenous IL-17A gene expression and suppressing differentiation of TH17 cells; stimulates T regulatory development