Learn More
Apexbio Technology LLC SR-18292-5MG
SDP
Catalog No. 5000568883
Encompass_Preferred
SR-18292 (CAS No 2095432-55-4) is a novel compound in the category of small molecule inhibitors originating from advanced synthetic chemistry processes It primarily targets cellular signaling pathways involved in inflammation and cancer specifically modulating protein kinases essential for cell proliferation In vitro SR-18292 demonstrates activity in the nanomolar to low micromolar range making it potent for biochemical assays Its typical applications include studies in various cell lines and animal models such as xenografts to assess anti-tumor efficacy and elucidate signaling mechanisms Additionally SR-18292 is utilized in drug screening programs to evaluate potential therapeutic interventions Experimental concentrations typically depend on the specific research design allowing for flexibility in research methodologies
Catalog No. 50-005-68883 Supplier Apexbio Technology LLC Supplier No. BA35825
SR-18292 (CAS No 2095432-55-4) is a novel compound in the category of small molecule inhibitors originating from advanced synthetic chemistry processes It primarily targets cellular signaling pathways involved in inflammation and cancer specifically modulating protein kinases essential for cell proliferation In vitro SR-18292 demonstrates activity in the nanomolar to low micromolar range making it potent for biochemical assays Its typical applications include studies in various cell lines and animal models such as xenografts to assess anti-tumor efficacy and elucidate signaling mechanisms Additionally SR-18292 is utilized in drug screening programs to evaluate potential therapeutic interventions Experimental concentrations typically depend on the specific research design allowing for flexibility in research methodologies