Learn More
Thermo Scientific SR-3000 Solvent Rack without Degassers for UltiMate™ 3000 Pumps
Catalog No. NC1661280 Shop All Thermo Scientific Products
Encompass
Securely organize up to nine 1L eluent bottles using the Thermo Scientific™ SR-3000 Solvent Rack without desgassers. Placed on top of your Thermo Scientific™ Dionex™ UltiMate™ 3000 system stack, the solvent rack without a degasser supports pumps with integrated degassers. It is recommended in combination with all UltiMate 3000 Quaternary RSLC, Standard, and Micro Pumps with internal degassers (LPG-3400RS/SD/AB/M/MB). Its compact design conserves valuable bench space. The tray is resistant to the most commonly used HPLC solvents and buffer solutions and enables easy cleaning and maintenance.
Catalog No. NC1661280 Supplier Thermo Scientific Supplier No. 5035.9200
Securely organize up to nine 1L eluent bottles using the Thermo Scientific™ SR-3000 Solvent Rack without desgassers. Placed on top of your Thermo Scientific™ Dionex™ UltiMate™ 3000 system stack, the solvent rack without a degasser supports pumps with integrated degassers. It is recommended in combination with all UltiMate 3000 Quaternary RSLC, Standard, and Micro Pumps with internal degassers (LPG-3400RS/SD/AB/M/MB). Its compact design conserves valuable bench space. The tray is resistant to the most commonly used HPLC solvents and buffer solutions and enables easy cleaning and maintenance.