Learn More
Medchemexpress LLC HY-13751 5mg Medchemexpress, SR-31747 CAS:132173-07-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY137515MG
Encompass_Preferred
Medchemexpress, HY-13751 5mg SR-31747 CAS:132173-07-0 SR-31747 is a sigma ligand with immunosuppressive and anti-inflammatory properties. SR-31747 blocks cell proliferation by inhibiting sterol isomerase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9888