Learn More
TARGETMOL CHEMICALS INC SR-318 10MG
SDP
Catalog No. 502581390
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR-318 potently inhibits the TNF-(alpha) release in whole blood (IC50 = 283 nM). SR-318 is a potent and highly selective inhibitor of p38 MAPK. The IC50 values are 5 nM 32 nM and 6.11 uM for p38(alpha) p38(beta) and p38(alpha)/(beta) respectively. purity: 99%
Catalog No. 50-258-1390 Supplier TARGETMOL CHEMICALS INC Supplier No. T1299710MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR-318 potently inhibits the TNF-(alpha) release in whole blood (IC50 = 283 nM). SR-318 is a potent and highly selective inhibitor of p38 MAPK. The IC50 values are 5 nM 32 nM and 6.11 uM for p38(alpha) p38(beta) and p38(alpha)/(beta) respectively. purity: 99%