Learn More
Medchemexpress LLC SR-4835 | 2387704-62-1 | 99.5% | 50 MG
SDP
Catalog No. 5000338416
Encompass_Preferred
SR-4835 is a potent, highly selective, and ATP-competitive dual inhibitor of CDK12/CDK13 (CDK12: IC50=99 nM, Kd=98 nM; CDK13: Kd=4.9 nM). It acts synergistically with DNA-damaging chemotherapy and PARP inhibitors, leading to cell death in triple-negative breast cancer (TNBC) cells.
  • Potent, highly selective, and ATP-competitive dual inhibitor of CDK12/CDK13
  • Inhibits CDK12 (IC50=99 nM, Kd=98 nM)
  • Inhibits CDK13 (Kd=4.9 nM)
  • Acts synergistically with DNA-damaging chemotherapy and PARP inhibitors
  • Leads to cell death in triple-negative breast cancer (TNBC) cells
  • Suppresses ATM and RAD51 protein levels
  • Triggers intronic polyadenylation site cleavage
  • Suppresses expression of core DNA damage response proteins
Catalog No. 50-003-38416 Supplier Medchemexpress LLC Supplier No. HY13025050MG
SR-4835 is a potent, highly selective, and ATP-competitive dual inhibitor of CDK12/CDK13 (CDK12: IC50=99 nM, Kd=98 nM; CDK13: Kd=4.9 nM). It acts synergistically with DNA-damaging chemotherapy and PARP inhibitors, leading to cell death in triple-negative breast cancer (TNBC) cells.
  • Potent, highly selective, and ATP-competitive dual inhibitor of CDK12/CDK13
  • Inhibits CDK12 (IC50=99 nM, Kd=98 nM)
  • Inhibits CDK13 (Kd=4.9 nM)
  • Acts synergistically with DNA-damaging chemotherapy and PARP inhibitors
  • Leads to cell death in triple-negative breast cancer (TNBC) cells
  • Suppresses ATM and RAD51 protein levels
  • Triggers intronic polyadenylation site cleavage
  • Suppresses expression of core DNA damage response proteins