Learn More
Medchemexpress LLC SR-4835 | 2387704-62-1 | 99.7% | 100 MG
SDP
Catalog No. 5000338316
Encompass_Preferred
SR-4835 is a potent, highly selective, and ATP-competitive dual inhibitor of CDK12/CDK13. It acts synergistically with DNA-damaging chemotherapy and PARP inhibitors to induce cell death in triple-negative breast cancer (TNBC) cells.
  • Potent and highly selective ATP-competitive dual inhibitor of CDK12/CDK13
  • Acts synergistically with DNA-damaging chemotherapy
  • Induces cell death in triple-negative breast cancer (TNBC) cells
  • Inhibits CDK12/CDK13, suppressing expression of core DNA damage response proteins
Catalog No. 50-003-38316 Supplier Medchemexpress LLC Supplier No. HY130250100MG
SR-4835 is a potent, highly selective, and ATP-competitive dual inhibitor of CDK12/CDK13. It acts synergistically with DNA-damaging chemotherapy and PARP inhibitors to induce cell death in triple-negative breast cancer (TNBC) cells.
  • Potent and highly selective ATP-competitive dual inhibitor of CDK12/CDK13
  • Acts synergistically with DNA-damaging chemotherapy
  • Induces cell death in triple-negative breast cancer (TNBC) cells
  • Inhibits CDK12/CDK13, suppressing expression of core DNA damage response proteins