Learn More
Selleck Chemical LLC SR-4835-5mg
SDP
Catalog No. 502389810
Encompass_Preferred
SR-4835 is a highly selective dual inhibitor of CDK12 and CDK13 with IC50 of 99 nM and Kd of 98 nM for CDK12 and IC50 of 4.9 nM for CDK13. SR-4835 disables triple-negative breast cancer (TNBC) cells. SR-4835 promotes synergy with DNA-damaging chemotherapy and PARP inhibitors.
Catalog No. 50-238-9810 Supplier Selleck Chemical LLC Supplier No. S88945MG
SR-4835 is a highly selective dual inhibitor of CDK12 and CDK13 with IC50 of 99 nM and Kd of 98 nM for CDK12 and IC50 of 4.9 nM for CDK13. SR-4835 disables triple-negative breast cancer (TNBC) cells. SR-4835 promotes synergy with DNA-damaging chemotherapy and PARP inhibitors.