Learn More
Medchemexpress LLC HY-130250 5mg Medchemexpress, SR-4835 CAS:2387704-62-1 Purity:>98%
SDP
Catalog No. 501969825
Encompass_Preferred
Medchemexpress, HY-130250 5mg SR-4835 CAS:2387704-62-1 SR-4835 is a potent, highly selective and ATP competitive dual inhibitor of CDK12/CDK13 (CDK12: IC50=99 nM, Kd=98 nM; CDK13: Kd=4.9 nM). SR-4835 acts in synergy with DNA-damaging chemotherapy and PARP inhibitors and provokes triple-negative breast cancer (TNBC) cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9825 Supplier Medchemexpress LLC Supplier No. HY1302505MG
Medchemexpress, HY-130250 5mg SR-4835 CAS:2387704-62-1 SR-4835 is a potent, highly selective and ATP competitive dual inhibitor of CDK12/CDK13 (CDK12: IC50=99 nM, Kd=98 nM; CDK13: Kd=4.9 nM). SR-4835 acts in synergy with DNA-damaging chemotherapy and PARP inhibitors and provokes triple-negative breast cancer (TNBC) cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.