Learn More
Eurofins Discoverx SR 49059, 10 mg. (92-1279)
Supplier:  Eurofins Discoverx 921279
Encompass
SR 49059 is an orally active synthetic small molecule that is a potent and selective inhibitor of the arginine vasopressin (AVP) V1a-receptor (Ki=1.1-6.3 nM). AVP is an important hormone for circulatory and water homeostasis, as well as renal hemodynamic alterations, water retention, and cardiac remodeling in congestive heart failure. SR 49059 shows promise in platelet aggregation inhibition and treatment of Raynaud's disease, dysmenorrhea, and tocolysis. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9796