Learn More
Cayman Chemical SR 58611A (hydrochloride), Purity: ≥98%, CAS: 121524-09-2, 5 mg
SDP
Catalog No. 502954087
Encompass_Preferred
A selective β3-AR agonist (EC50 = 3.5 nM in rat colon); minimally active against β1-ARs in rat uterus (EC50 = 499 nM) and inactive against β2-ARs in guinea pig trachea and atrium (EC50s = >10,000 and >30,000 nM, respectively); activates brown fat metabolism through activation of adenylyl cyclase activity and glycerol release in brown adipocytes (EC50s = 20 and 11 nM, respectively); reduces hypothermia produced by apomorphine and reserpine; potentiates toxicity produced by yohimbine; reduces number of escape failures in a learned helplessness model of antidepressant-like activity in rats
Catalog No. 50-295-4087 Supplier Cayman Chemical Supplier No. 119545 MG
A selective β3-AR agonist (EC50 = 3.5 nM in rat colon); minimally active against β1-ARs in rat uterus (EC50 = 499 nM) and inactive against β2-ARs in guinea pig trachea and atrium (EC50s = >10,000 and >30,000 nM, respectively); activates brown fat metabolism through activation of adenylyl cyclase activity and glycerol release in brown adipocytes (EC50s = 20 and 11 nM, respectively); reduces hypothermia produced by apomorphine and reserpine; potentiates toxicity produced by yohimbine; reduces number of escape failures in a learned helplessness model of antidepressant-like activity in rats