Learn More
Cayman Chemical SR 59230A (hydrochloride), Purity: ≥99%, CAS: 1135278-41-9, 10 mg
SDP
Catalog No. 502993978
Encompass_Preferred
A β3-AR antagonist in animal models; less selective for human β-AR subtypes (Kis = 16.4, 61.9, and 122 nM for β1-, β2-, and β3-AR, respectively); blocks MDMA-induced hyperthermia at low and high concentrations with some α1-AR antagonist properties
Catalog No. 50-299-3978 Supplier Cayman Chemical Supplier No. 2140710 MG
A β3-AR antagonist in animal models; less selective for human β-AR subtypes (Kis = 16.4, 61.9, and 122 nM for β1-, β2-, and β3-AR, respectively); blocks MDMA-induced hyperthermia at low and high concentrations with some α1-AR antagonist properties