Learn More
Medchemexpress LLC HY-131454 10mg , SR-717 CAS:2375421-09-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13145410MG
Encompass_Preferred
HY-131454 10mg SR-717 CAS: 2375421-09-1 SR-717 is a non-nucleotide STING agonist with EC50s of 2.1 μM and 2.2 μM in ISG-THP1 (WT) and ISG-THP1 cGAS KO (cGAS KO) cell lines, respectively. SR-717 is a stable cyclic guanosine monophosphate-adenosine monophosphate (cGAMP) mimetic. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9096