Learn More
Cayman Chemical SR 717 (lithium salt), Purity: ≥98%, CAS: 2375421-09-1, 25 mg
SDP
Catalog No. 502990707
Encompass_Preferred
A STING agonist (EC50 = 2.1 µM in a reporter assay); selective for STING over a panel of 468 kinases; increases plasma IFN-β and IL-6 levels in mice at 30 mg/kg; increases the intratumor frequency of CD69+CD8+ T cells, reduces tumor growth, and increases survival in a B16/F10 murine melanoma model
Catalog No. 50-299-0707 Supplier Cayman Chemical Supplier No. 3175925 MG
A STING agonist (EC50 = 2.1 µM in a reporter assay); selective for STING over a panel of 468 kinases; increases plasma IFN-β and IL-6 levels in mice at 30 mg/kg; increases the intratumor frequency of CD69+CD8+ T cells, reduces tumor growth, and increases survival in a B16/F10 murine melanoma model